Benzamidin Hydrochlorid Hydrat, (H₂O 10-14%) 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Phenylamidin Hydrochlorid Hydrat, Benzoesäureamidin Hydrochlorid Hydrat, Benzamidiniumchlorid Hydrat
J62823.06
J62823.09
J62823.06EA
37.8
EUR
J62823.06
J62823.09
Benzamidin Hydrochlorid Hydrat, (H₂O 10-14%) 99%
Benzamidin Hydrochlorid Hydrat
Purity: 99%
CAS No.: 206752-36-5
Molecular Formula: C7H8N26.HCl.xH2O
Formula Weight: 156.62
Storage Temperature: 2°C to 8°C
Physical Form: Powder or beads
Melting Point: 80-85°
Beilstein Reference: 3594959
MDL No.: MFCD00013025
Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases
Formel:
C₇H₉ClN₂ Molecular Weight: 174,63 g/mol Schmelzpunkt: 80…85 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00066285 CAS-Nummer: 1670-14-0 EINECS: 216-795-4 |