Genistein ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-on, 4',5,7-Trihydroxyisoflavon, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyron
4',5,7-Trihydroxyisoflavone is used to inhibitor of tyrosine protein kinase. Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
Purity: 99+%
CAS No.: 446-72-0
Molecular Formula: C15H10O5
Formula Weight: 270.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Melting Point: ca 300°
Beilstein Reference: 263823
Merck Reference: 14,4391
MDL No.: MFCD00016952
Inhibitor of tyrosine protein kinase
Formel:
C₁₅H₁₀O₅ Schmelzpunkt: ∼ 300 °C |
MDL:
MFCD00016952 CAS-Nummer: 446-72-0 EINECS: 207-174-9 Merck Index: 14,4391 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...