Tyramin ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: 2-(4-Hydroxyphenyl)ethylamin, 4-Hydroxyphenethylamim, 4-(2-Aminoethyl)phenol
J60990.03 J60990.06 J60990.14
J60990.03EA 30.4 EUR
J60990.03 J60990.06 J60990.14
Tyramin ≥98%
Tyramin
Purity: 98+% CAS No.: 51-67-2 Molecular Formula: C8H11NO Formula Weight: 137.18 Physical Form: Powder Appearance: Off-white to yellow Melting Point: 160-162° Boiling Point: 175-181°/8mm Hg Beilstein Reference: 1099914 Merck Reference: 14,9835 MDL No.: MFCD00008193
Reagent for fluorescence enzyme immunoassay
Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Siedepunkt: 175…181 °C (8 mmHg)
Schmelzpunkt: 160…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008193
CAS-Nummer: 51-67-2
EINECS: 200-115-8
Merck Index: 14,9835