2-Chlor-6-nitrobenzothiazol 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Chloro-6-nitrobenzo[d]thiazole, 2-Chloro-6-nitro-1,3-benzothiazole

H35525.03 H35525.09
H35525.03EA 38.3 EUR
H35525.03 H35525.09
2-Chlor-6-nitrobenzothiazol 95%
2-Chloro-6-nitrobenzothiazole
Formel: C₇H₃ClN₂O₂S
Molecular Weight: 214,63 g/mol
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022852
CAS-Nummer: 2407-11-6
EINECS: 219-302-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR