8-Amino-2-naphthalinsulfonsäure 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-Naphthylamine-7-sulphonic acid, 1,7-Cleve's acid
J65304.14
J65304.22
J65304.14EA
68.8
EUR
J65304.14
J65304.22
8-Amino-2-naphthalinsulfonsäure 95%
8-Amino-2-naphthalenesulphonic acid
Formel:
C₁₀H₉NO₃S Molecular Weight: 223,25 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004032 CAS-Nummer: 119-28-8 EINECS: 204-311-4 UN: 2585 ADR: 8,III Merck Index: 142,351 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...