8-Anilino-1-naphthalinsulfonsäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid
J65332.06
J65332.14
J65332.22
J65332.06EA
42.5
EUR
J65332.06
J65332.14
J65332.22
8-Anilino-1-naphthalinsulfonsäure 95%
8-Anilino-1-naphthalinsulfonsäure
Formel:
C₁₆H₁₃NO₃S Molecular Weight: 299,35 g/mol Schmelzpunkt: 215…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003998 CAS-Nummer: 82-76-8 EINECS: 201-438-7 Merck Index: 14,661 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...