TAPS-Natriumsalz (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure Natriumsalz) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
TAPS sodium salt, Sodium 3-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)propane-1-sulphonate
J61189.14
J61189.22
J61189.14EA
28
EUR
J61189.14
J61189.22
TAPS-Natriumsalz (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure Natriumsalz) ≥98%
TAPS-Natriumsalz (3-(Tris(hydroxymethyl)methylamino)propansulfonsäure Natriumsalz)
Purity: 98%
CAS No.: 91000-53-2
Molecular Formula: C7H16NNaO6S
Formula Weight: 265.26
Physical Form: Powder
Appearance: White
MDL No.: MFCD00069903
Zwitterionic buffer
Formel:
C₇H₁₇NO₆S Molecular Weight: 265.26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069903 CAS-Nummer: 91000-53-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...