2-Chlor-3-methyl-5-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-5-nitro-3-picoline

PY-1664-25G PY-1664-100G
COBBPY-1664-25GEA 90.7 EUR
COBBPY-1664-25G COBBPY-1664-100G
2-Chlor-3-methyl-5-nitropyridin
2-Chloro-3-methyl-5-nitropyridine
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.57 g/mol
MDL: MFCD00173686
CAS-Nummer: 22280-56-4
EINECS: 244-889-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR