Ketoprofen

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(3-Benzoylphenyl)propionic acid, 2-(3-Benzoylphenyl)propanoic acid

J62702.06 J62702.14
J62702.06EA 85.2 EUR
J62702.06 J62702.14
Ketoprofen
Ketoprofen
Formel: C₁₆H₁₄O₃
Molecular Weight: 254,29 g/mol
Schmelzpunkt: 92…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055790
CAS-Nummer: 22071-15-4
EINECS: 244-759-8
UN: 2811
ADR: 6.1,III
Merck Index: 14,5305

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR