Guanosin-5'-diphosphat Dinatriumsalz
Lieferant: Thermo Scientific
Synonyme:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate, disodium salt, Guanosine 5'-diphosphate disodium salt
J61646.MC
J61646.ME
J61646.MCEA
119
EUR
J61646.MC
J61646.ME
Guanosin-5'-diphosphat Dinatriumsalz
Guanosin-5'-diphosphat Dinatriumsalz
CAS No.: 7415-69-2
Molecular Formula: C10H13N5Na2O11P2
Formula Weight: 487.17
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00084665
Formel:
C₁₀H₁₃N₅Na₂O₁₁P₂ Molecular Weight: 487,17 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00070121 CAS-Nummer: 7415-69-2 EINECS: 231-026-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...