Guanosin-5'-triphosphat Dinatriumsalz
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Guanosine-5'-triphosphate disodium salt, GTP disodium salt, Guanosine-5-triphosphoric aicd disodium salt
J61414.MD
J61414.ME
J61414.03
J61414.MDEA
245
EUR
J61414.MD
J61414.ME
J61414.03
Guanosin-5'-triphosphat Dinatriumsalz
Guanosin-5'-triphosphat Dinatriumsalz
CAS No.: 56001-37-7
Molecular Formula: C10H14N5Na2O14P3
Formula Weight: 567.15
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 4113439
MDL No.: MFCD00083629
Formel:
C₁₀H₁₄N₅Na₂O₁₄P₃ Molecular Weight: 567.147 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD03410297 CAS-Nummer: 56001-37-7 EINECS: 259-940-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...