2'-Deoxyinosin
Lieferant: Thermo Scientific
Synonyme:
9-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-9H-purin-6-ol
J61411.09
J61411.14
J61411.09EA
160
EUR
J61411.09
J61411.14
2'-Deoxyinosin
2'-Deoxyinosin
CAS No.: 890-38-0
Molecular Formula: C10H12N4O4
Formula Weight: 252.23
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline powder
Appearance: White to off-white
Melting Point:
Formel:
C₁₀H₁₂N₄O₄ Molecular Weight: 252,23 g/mol Schmelzpunkt: >250 °C |
MDL:
MFCD00005762 CAS-Nummer: 890-38-0 EINECS: 212-964-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...