2'-Desoxyadenosin ≥99%
Lieferant: Thermo Scientific
Synonyme:
(2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol
J63886.18
J63886.22
J63886.18EA
258
EUR
J63886.18
J63886.22
2'-Desoxyadenosin ≥99%
2'-Desoxyadenosin
Purity: 99%
CAS No.: 958-09-8
Molecular Formula: C10H13N5O3
Formula Weight: 251.24
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 50mg/ml
Optical Rotation: -25° (c=0.5 in water)
MDL No.: MFCD00056003
Formel:
C₁₀H₁₃N₅O₃ Molecular Weight: 251,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00056003 CAS-Nummer: 958-09-8 EINECS: 213-488-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...