Octyl-β-D-thioglucopyranosid ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside
Purity: 98+%
CAS No.: 85618-21-9
Molecular Formula: C14H28O5S
Formula Weight: 308.44
Physical Form: Crystalline powder
Appearance: White to off-white
Melting Point: 128°
Solubility: Soluble in ethanol, water or methanol
Beilstein Reference: 4182783
MDL No.: MFCD00012189
Non-ionic detergent used for the solubilization of membrane proteins
Formel:
C₁₄H₂₈O₅S Molecular Weight: 308.44 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012189 CAS-Nummer: 85618-21-9 |