Nalidixinsäure Natriumsalz

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Sodium 1-ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate

J63550.06 J63550.14 J63550.22 J63550.36
J63550.06EA 41.5 EUR
J63550.06 J63550.14 J63550.22 J63550.36
Nalidixinsäure Natriumsalz
Nalidixinsäure Natriumsalz

CAS No.: 3374-05-8 Molecular Formula: C12H11N2NaO3 Formula Weight: 254.22 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white to pale yellow Melting Point: 280° Solubility: Soluble at 50mg/ml in water Beilstein Reference: 3580062 MDL No.: MFCD00064376


Inhibitor of bacterial DNA gyrase. An antibacterial agent

Formel: C₁₂H₁₁N₂NaO₃
Schmelzpunkt: >280 °C
MDL: MFCD00064376
CAS-Nummer: 3374-05-8
EINECS: 222-159-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR