2-Fluor-4-(trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-4-(trifluoromethyl)benzeneboronic acid
BB-3559-5G
BB-3559-1G
COBBBB-3559-5GEA
147
EUR
COBBBB-3559-5G
COBBBB-3559-1G
2-Fluor-4-(trifluormethyl)phenylboronsäure
2-Fluor-4-(trifluormethyl)phenylboronsäure
Formel:
C₇H₅BF₄O₂ Molecular Weight: 207.92 g/mol |
MDL:
MFCD03094995 CAS-Nummer: 503309-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...