3,5-Dibrom-α,α,α-trifluortoluol
Lieferant: COMBI-BLOCKS
Synonyme:
3,5-Dibromobenzotrifluoride, 1,3-Dibromo-5-(trifluoromethyl)benzene
OT-0431-5G
OT-0431-25G
OT-0431-1G
COBBOT-0431-5GEA
75.9
EUR
COBBOT-0431-5G
COBBOT-0431-25G
COBBOT-0431-1G
3,5-Dibrom-α,α,α-trifluortoluol
3,5-Dibrom-α,α,α-trifluortoluol
Formel:
C₇H₃Br₂F₃ Molecular Weight: 303.90 g/mol |
MDL:
MFCD00526219 CAS-Nummer: 401-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...