Methyl-3-amino-2-benzo[b]thiophencarboxylat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 3-aminobenzo[b]thiophene-2-carboxylate
H33956.03
H33956.06
H33956.03EA
37.9
EUR
H33956.03
H33956.06
Methyl-3-amino-2-benzo[b]thiophencarboxylat 97%
Methyl-3-amino-2-benzo[b]thiophencarboxylat
Purity: 97%
CAS No.: 35212-85-2
Molecular Formula: C10H9NO2S
Formula Weight: 207.25
Melting Point: 107-108°
MDL No.: MFCD00206744
Formel:
C₁₀H₉NO₂S Schmelzpunkt: 107…108 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00206744 CAS-Nummer: 35212-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...