4-Brom-2-(morpholin-4-ylsulfonyl)anisol

Lieferant: COMBI-BLOCKS

Synonyme: 4-((5-Bromo-2-methoxyphenyl)sulphonyl)morpholine

OT-0197-5G OT-0197-1G
COBBOT-0197-5GEA 692 EUR
COBBOT-0197-5G COBBOT-0197-1G
4-Brom-2-(morpholin-4-ylsulfonyl)anisol
4-Brom-2-(morpholin-4-ylsulfonyl)anisol
Formel: C₁₁H₁₄BrNO₄S
Molecular Weight: 336,2000 g/mol
MDL: MFCD02043681
CAS-Nummer: 325809-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR