Pentafluorphenyl-1-ethylensulfonat

Lieferant: BIONET RESEARCH

4Y-0324.10 4Y-0324.1 4Y-0324.500 4Y-0324.5
BIOT4Y-0324.10EA 0 EUR
BIOT4Y-0324.10 BIOT4Y-0324.1 BIOT4Y-0324.500 BIOT4Y-0324.5
Pentafluorphenyl-1-ethylensulfonat
Pentafluorphenyl-1-ethylensulfonat
Formel: C₈H₃F₅O₃S
MDL: MFCD06659554
CAS-Nummer: 452905-58-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR