Pentafluorphenyl-2,3-dichlorbenzolsulfonat

Lieferant: BIONET RESEARCH

1Y-0264.500 1Y-0264.10 1Y-0264.1 1Y-0264.5
BIOT1Y-0264.500EA 0 EUR
BIOT1Y-0264.500 BIOT1Y-0264.10 BIOT1Y-0264.1 BIOT1Y-0264.5
Pentafluorphenyl-2,3-dichlorbenzolsulfonat
Pentafluorphenyl-2,3-dichlorbenzolsulfonat
Formel: C₁₂H₃Cl₂F₅O₃S
MDL: MFCD06659539
CAS-Nummer: 885949-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR