Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Lieferant: BIONET RESEARCH
1Y-0271.1
1Y-0271.10
1Y-0271.5
1Y-0271.500
BIOT1Y-0271.1EA
0
EUR
BIOT1Y-0271.1
BIOT1Y-0271.10
BIOT1Y-0271.5
BIOT1Y-0271.500
Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Pentafluorphenyl-2,4-dichlorbenzolsulfonat
Formel:
C₁₂H₃Cl₂F₅O₃S |
MDL:
MFCD06659543 CAS-Nummer: 885949-57-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...