Pentafluorphenyl-2-chlorbenzolsulfonat

Lieferant: BIONET RESEARCH

1Y-0267.1 1Y-0267.10 1Y-0267.5 1Y-0267.500
BIOT1Y-0267.1EA 0 EUR
BIOT1Y-0267.1 BIOT1Y-0267.10 BIOT1Y-0267.5 BIOT1Y-0267.500
Pentafluorphenyl-2-chlorbenzolsulfonat
Pentafluorphenyl-2-chlorbenzolsulfonat
Formel: C₁₂H₄ClF₅O₃S
MDL: MFCD06659541
CAS-Nummer: 885949-55-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR