Pentafluorphenyl-4-cyanbenzolsulfonat
Lieferant: BIONET RESEARCH
1Y-0268.5
1Y-0268.10
1Y-0268.500
1Y-0268.1
BIOT1Y-0268.5EA
0
EUR
BIOT1Y-0268.5
BIOT1Y-0268.10
BIOT1Y-0268.500
BIOT1Y-0268.1
Pentafluorphenyl-4-cyanbenzolsulfonat
Pentafluorphenyl-4-cyanbenzolsulfonat
Formel:
C₁₃H₄F₅NO₃S |
MDL:
MFCD06659542 CAS-Nummer: 885949-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...