2-Brom-5-fluorisonicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Bromo-5-fluoro-4-pyridinecarboxylic acid, 2-Bromo-5-fluoropyridine-4-carboxylic acid

OT-8080-1G OT-8080-25G OT-8080-5G
COBBOT-8080-1GEA 31 EUR
COBBOT-8080-1G COBBOT-8080-25G COBBOT-8080-5G
2-Brom-5-fluorisonicotinsäure
2-Brom-5-fluorisonicotinsäure
Formel: C₆H₃BrFNO₂
Molecular Weight: 219.9959 g/mol
MDL: MFCD09261246
CAS-Nummer: 885588-12-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR