Pentafluorophenyl-5-brom-2-thiophensulfonat

Lieferant: BIONET RESEARCH

4Y-0246.1 4Y-0246.10 4Y-0246.5 4Y-0246.500
BIOT4Y-0246.1EA 0 EUR
BIOT4Y-0246.1 BIOT4Y-0246.10 BIOT4Y-0246.5 BIOT4Y-0246.500
Pentafluorophenyl-5-brom-2-thiophensulfonat
Pentafluorophenyl-5-brom-2-thiophensulfonat
Formel: C₁₀H₂BrF₅O₃S₂
MDL: MFCD06659553
CAS-Nummer: 885949-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR