2'-Brom-4'-chloracetanilid

Lieferant: COMBI-BLOCKS

AN-1373-5G AN-1373-1G
COBBAN-1373-5GEA 884 EUR
COBBAN-1373-5G COBBAN-1373-1G
2'-Brom-4'-chloracetanilid
2'-Brom-4'-chloracetanilid
Formel: C₈H₇BrClNO
MDL: MFCD00040851
CAS-Nummer: 57045-85-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR