2-Methoxythioanisol

Lieferant: FLUOROCHEM

Synonyme: 2-(Methylsulphanyl)anisole

008048-5G 008048-25G
FLUO008048-5GEA 48.8 EUR
FLUO008048-5G FLUO008048-25G
2-Methoxythioanisol
2-Methoxythioanisol
Formel: C₈H₁₀OS
MDL: MFCD00792424
CAS-Nummer: 2388-73-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR