N-[2-Nitro-4-(trifluormethyl)phenyl]pyrrolidin

Lieferant: Thermo Scientific

BTB01877.1 BTB01877.10 BTB01877.50
MAYBBTB01877.1EA 0 EUR
MAYBBTB01877.1 MAYBBTB01877.10 MAYBBTB01877.50
N-[2-Nitro-4-(trifluormethyl)phenyl]pyrrolidin
N-[2-Nitro-4-(trifluormethyl)phenyl]pyrrolidin
Formel: C₁₁H₁₁F₃N₂O₂
MDL: MFCD00052886
CAS-Nummer: 40832-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR