4-(Methylthio)benzylamin

Lieferant: FLUOROCHEM

Synonyme: (4-(Methylthio)phenyl)methanamine

009292-5G 009292-1G
FLUO009292-5GEA 282 EUR
FLUO009292-5G FLUO009292-1G
4-(Methylthio)benzylamin
4-(Methylthio)benzylamin
Formel: C₈H₁₁NS
Molecular Weight: 153,2500 g/mol
MDL: MFCD01310831
CAS-Nummer: 83171-39-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR