4'-Fluor-2'-methoxyacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(4-Fluoro-2-methoxyphenyl)ethanone

JFD01249.250 JFD01249.1
CHMPJFD01249.250EA 0 EUR
CHMPJFD01249.250 MAYBJFD01249.1
4'-Fluor-2'-methoxyacetophenon
4'-Fluor-2'-methoxyacetophenon
Formel: C₉H₉FO₂
Molecular Weight: 168,17 g/mol
MDL: MFCD00061143
CAS-Nummer: 51788-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR