2'-Chlor-6'-fluor-3'-methylacetophenon
Lieferant: FLUOROCHEM
Synonyme:
1-(2-Chloro-6-fluoro-3-methylphenyl)ethanone
005638-5G
005638-1G
FLUO005638-5GEA
296
EUR
FLUO005638-5G
FLUO005638-1G
2'-Chlor-6'-fluor-3'-methylacetophenon
2'-Chlor-6'-fluor-3'-methylacetophenon
Formel:
C₉H₈ClFO |
MDL:
MFCD01631368 CAS-Nummer: 261762-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...