3-Iod-1H-pyrazol
Lieferant: COMBI-BLOCKS
Synonyme:
3-Iodopyrazole
HI-1602-5G
HI-1602-1G
COBBHI-1602-5GEA
157
EUR
COBBHI-1602-5G
COBBHI-1602-1G
3-Iod-1H-pyrazol
3-Iodo-1H-pyrazole
Formel:
C₃H₃IN₂ Molecular Weight: 193,98 g/mol |
MDL:
MFCD12022324 CAS-Nummer: 4522-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...