3-Formyl-4-methoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Formyl-4-methoxybenzeneboronic acid
L17850.03
L17850.06
L17850.03EA
25.9
EUR
L17850.03
L17850.06
3-Formyl-4-methoxyphenylboronsäure ≥98%
3-Formyl-4-methoxyphenylboronsäure
Purity: 98%
CAS No.: 121124-97-8
Molecular Formula: C8H9BO4
Formula Weight: 179.97
Melting Point: ca 240° dec.
Beilstein Reference: 7423998
Formel:
C₈H₉BO₄ Molecular Weight: 179,97 g/mol Schmelzpunkt: ∼ 240 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093661 CAS-Nummer: 121124-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...