3-Formyl-4-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Formyl-4-methoxybenzeneboronic acid

BB-2301-1G BB-2301-5G
COBBBB-2301-1GEA 26.2 EUR
COBBBB-2301-1G COBBBB-2301-5G
3-Formyl-4-methoxyphenylboronsäure
3-Formyl-4-methoxyphenylboronsäure
Formel: C₈H₉BO₄
Molecular Weight: 179.97 g/mol
MDL: MFCD02093661
CAS-Nummer: 121124-97-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR