3-Formyl-4-methoxyphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Formyl-4-methoxybenzeneboronic acid

021614-5G 021614-2G
FLUO021614-5GEA 97.2 EUR
FLUO021614-5G FLUO021614-2G
3-Formyl-4-methoxyphenylboronsäure
3-Formyl-4-methoxyphenylboronsäure
Formel: C₈H₉BO₄
MDL: MFCD02093661
CAS-Nummer: 121124-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR