5-Formyl-2-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Formyl-2-methoxybenzeneboronic acid, (5-Formyl-2-methoxyphenyl)boronic acid
BB-2302-5G
BB-2302-1G
COBBBB-2302-5GEA
49.7
EUR
COBBBB-2302-5G
COBBBB-2302-1G
5-Formyl-2-methoxyphenylboronsäure
5-Formyl-2-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179,9700 g/mol |
MDL:
MFCD01319044 CAS-Nummer: 127972-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...