5-Formyl-2-methoxyphenylboronsäure
Lieferant: FLUOROCHEM
Synonyme:
5-Formyl-2-methoxybenzeneboronic acid, (5-Formyl-2-methoxyphenyl)boronic acid
011045-5G
011045-25G
FLUO011045-5GEA
76.4
EUR
FLUO011045-5G
FLUO011045-25G
5-Formyl-2-methoxyphenylboronsäure
5-Formyl-2-methoxyphenylboronsäure
Formel:
C₈H₉BO₄ Molecular Weight: 179.9700 g/mol |
MDL:
MFCD01319044 CAS-Nummer: 127972-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...