5-Brom-2-(trifluormethoxy)anilin
Lieferant: COMBI-BLOCKS
Synonyme:
3-Amino-4-(trifluoromethoxy)bromobenzene
OS-7531-5G
OS-7531-25G
OS-7531-100G
COBBOS-7531-5GEA
26.2
EUR
COBBOS-7531-5G
COBBOS-7531-25G
COBBOS-7531-100G
5-Brom-2-(trifluormethoxy)anilin
5-Brom-2-(trifluormethoxy)anilin
Formel:
C₇H₅BrF₃NO Molecular Weight: 256.0200 g/mol |
MDL:
MFCD04039242 CAS-Nummer: 886762-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...