4-Methylnicotinamid

Lieferant: CHEMPUR

Synonyme: 4-Methyl-3-pyridinecarboxamide

RF04512.250 RF04512.1
CHMPRF04512.250EA 0 EUR
CHMPRF04512.250 CHMPRF04512.1
4-Methylnicotinamid
4-Methylnicotinamid
Formel: C₇H₈N₂O
Molecular Weight: 136.1512 g/mol
MDL: MFCD00234322
CAS-Nummer: 7250-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR