5-Methylnicotinamid ≥97%

Lieferant: Thermo Scientific
Achtung

L18970.03 L18970.06
L18970.03EA 71.2 EUR
L18970.03 L18970.06
5-Methylnicotinamid ≥97%
5-Methylnicotinamid

Purity: 97% CAS No.: 70-57-5 Molecular Formula: C7H8N2O Formula Weight: 136.15 Melting Point: 166-168° Beilstein Reference: 4962976

Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Schmelzpunkt: 166…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05664041
CAS-Nummer: 70-57-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR