4-(2-Aminoethyl)tetrahydropyran

Lieferant: COMBI-BLOCKS

Synonyme: 4-Aminoethyl-tetrahydropyrane, 4-(2-Aminoethyl)tetrahydro-2H-pyran

AM-1001-5G AM-1001-1G
COBBAM-1001-5GEA 1270 EUR
COBBAM-1001-5G COBBAM-1001-1G
4-(2-Aminoethyl)tetrahydropyran
4-(2-Aminoethyl)tetrahydropyran
Formel: C₇H₁₅NO
Molecular Weight: 129.2001 g/mol
MDL: MFCD02179433
CAS-Nummer: 65412-03-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR