4-(2-Aminoethyl)tetrahydropyran Hydrochlorid
Lieferant: COMBI-BLOCKS
Synonyme:
2-(Tetrahydro-2H-pyran-4-yl)ethanamine hydrochloride, 4-(2-Aminoethyl)tetrahydropyran, HCl
AM-1011-1G
AM-1011-5G
COBBAM-1011-1GEA
236
EUR
COBBAM-1011-1G
COBBAM-1011-5G
4-(2-Aminoethyl)tetrahydropyran Hydrochlorid
4-(2-Aminoethyl)tetrahydropyran Hydrochlorid
Formel:
C₇H₁₆ClNO Molecular Weight: 165.66 g/mol |
MDL:
MFCD03411927 CAS-Nummer: 389621-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...