2-Brom-5'-chlor-2'-hydroxyacetophenone, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

663689-1G 663689-5G
SIAL663689-1GEA 38 EUR
SIAL663689-1G SIAL663689-5G
2-Brom-5'-chlor-2'-hydroxyacetophenone, Sigma-Aldrich®
2-Brom-5'-chlor-2'-hydroxyacetophenone
Formel: C₈H₆BrClO₂
MDL: MFCD03789076
CAS-Nummer: 52727-99-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR