2-Brom-4'-(difluormethoxy)acetophenon
Lieferant: FLUOROCHEM
Synonyme:
4-(Difluoromethoxy)phenacyl bromide
004841-1G
004841-5G
FLUO004841-1GEA
273
EUR
FLUO004841-1G
FLUO004841-5G
2-Brom-4'-(difluormethoxy)acetophenon
2-Brom-4'-(difluormethoxy)acetophenon
Formel:
C₉H₇BrF₂O₂ |
MDL:
MFCD00052200 CAS-Nummer: 141134-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...