(-)-Menthylchlorformiat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1R)-(-)-Menthyl chloroformate, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl carbonochloridate

M-Clarity™ Qualitätsstufe = MQ100

245305-100G 245305-25G
SIAL245305-100GEA 454 EUR
SIAL245305-100G SIAL245305-25G
(-)-Menthylchlorformiat, Sigma-Aldrich®
(-)-Menthylchlorformiat
Formel: C₁₁H₁₉ClO₂
Molecular Weight: 218,73 g/mol
MDL: MFCD00009694
CAS-Nummer: 14602-86-9
UN: 3277
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR