(+)-Menthylchlorformiat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (1S,2R,5S)-2-Isopropyl-5-methylcyclohexyl carbonochloridate, (1S)-(+)-Menthyl chloroformate

M-Clarity™ Qualitätsstufe = MQ100

378712-25ML 378712-5ML
SIAL378712-25MLEA 215 EUR
SIAL378712-25ML SIAL378712-5ML
(+)-Menthylchlorformiat, Sigma-Aldrich®
(+)-Menthylchlorformiat
Formel: C₁₁H₁₉ClO₂
Molecular Weight: 218,720 g/mol
MDL: MFCD00134483
CAS-Nummer: 7635-54-3
UN: 3277
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR