Iminodiacetonitril 95%
Lieferant: Thermo Scientific
|
Gefahr
|
B23660.0E
B23660.22
B23660.36
B23660.0EEA
433
EUR
B23660.0E
B23660.22
B23660.36
Iminodiacetonitril 95%
Iminodiacetonitril
Purity: 95%
CAS No.: 628-87-5
Molecular Formula: HN(CH2CN)2
Formula Weight: 95.11
Melting Point: 75-79°
Beilstein Reference: 773803
Formel:
C₄H₅N₃ Molecular Weight: 95,11 g/mol Schmelzpunkt: 75…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001887 CAS-Nummer: 628-87-5 EINECS: 211-058-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...