Di-tert-butyliminodicarboxylat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Di-(tert-butyl) imidodicarbonate
L20325.06
L20325.14
L20325.06EA
91.4
EUR
L20325.06
L20325.14
Di-tert-butyliminodicarboxylat 98%
Di-tert-butyliminodicarboxylat
Purity: 98%
CAS No.: 51779-32-9
Molecular Formula: C10H19NO4
Formula Weight: 217.27
Melting Point: 114-119°
Beilstein Reference: 1911172
Formel:
C₁₀H₁₉NO₄ Molecular Weight: 217.27 g/mol Schmelzpunkt: 114…119 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00043309 CAS-Nummer: 51779-32-9 |