N-Boc-glycin (Boc-Gly-OH)
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Boc-Gly-OH
4001229.0100
4001229.0500
4001229.1000
BACE4001229.0100EA
0
EUR
BACE4001229.0100
BACE4001229.0500
BACE4001229.1000
N-Boc-glycin (Boc-Gly-OH)
N-Boc-glycin (Boc-Gly-OH)
Formel:
C₇H₁₃NO₄ Molecular Weight: 175,2 g/mol |
MDL:
MFCD00002690 CAS-Nummer: 4530-20-5 EINECS: 224-864-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...